siRNA Details
Gene Symbol: C14orf159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102366.2 | NP_001095836.1 | 6 | 947 | ||
NM_001102367.2 | NP_001095837.1 | 5 | 713 | |||
NM_001102368.2 | NP_001095838.1 | 4 | 531 | |||
NM_001102369.2 | NP_001095839.1 | 6 | 702 | |||
NM_001286470.1 | NP_001273399.1 | 7 | 1226 | |||
NM_001286471.1 | NP_001273400.1 | 4 | 782 | |||
NM_001286472.1 | NP_001273401.1 | 6 | 702 | |||
NM_001286473.1 | NP_001273402.1 | 6 | 702 | |||
NM_024952.7 | NP_079228.4 | 4 | 776 | |||
GenBank mRNA | AB210031.1 | - | 4 | 432 | ||
AK022564.1 | - | 4 | 347 | |||
AK023200.1 | - | 6 | 500 | |||
AK097294.1 | - | 4 | 481 | |||
AK098752.1 | - | 4 | 482 | |||
AK123532.1 | - | 7 | 869 | |||
AK308857.1 | - | 3 | 393 | |||
AY358516.1 | - | 4 | 782 | |||
BC009182.2 | - | 6 | 628 | |||
BC010614.2 | - | 5 | 662 | 4157432 | ||
BC065558.1 | - | 6 | 570 | 6138321 | ||
BX161393.1 | - | 4 | 460 | |||
BX247989.1 | - | 6 | 630 | |||
BX248060.1 | - | 4 | 585 | |||
BX248294.1 | - | 2 | 133 | |||
BX248782.1 | - | 5 | 481 | |||
BX537970.1 | - | 4 | 478 | |||
CR457289.1 | - | 2 | 179 | |||
DA296867.1 | - | 4 | 531 |
