siRNA Details
Gene Symbol: CALU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130674.2 | NP_001124146.1 | 7 | 5248 | ||
NM_001199671.1 | NP_001186600.1 | 8 | 6063 | |||
NM_001199672.1 | NP_001186601.1 | 8 | 6063 | |||
NM_001199673.1 | NP_001186602.1 | 6 | 5048 | |||
NM_001219.4 | NP_001210.1 | 7 | 5248 | |||
NR_074086.1 | 5 | 4822 |
Gene Symbol: FLJ11710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021772.1 | - | 1 | 799 |
Gene Symbol: LINC01194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033383.1 | 3 | 292 | |||
GenBank mRNA | AY328031.1 | - | 3 | 292 | ||
AY328032.1 | - | 3 | 292 | |||
AY328033.1 | - | 3 | 292 | |||
AY330599.1 | - | 2 | 210 |
Gene Symbol: RAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127392.1 | - | 2 | 548 |
Gene Symbol: ZNF431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319126.1 | NP_001306055.1 | 4, 5 | 501 | ||
GenBank mRNA | BX647597.1 | - | 4, 5 | 487 |
Gene Symbol: ZNF714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117086.1 | 4, 5 | 525 | |||
GenBank mRNA | BC022527.1 | - | 4, 5 | 469 | ||
BC037782.1 | - | 4, 5 | 474 | 4797729 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
