siRNA Details
Gene Symbol: FBXO38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271723.1 | NP_001258652.1 | 3 | 359 | ||
NM_030793.4 | NP_110420.3 | 3 | 359 | |||
XM_005268513.1 | XP_005268570.1 | 3 | 348 | |||
XM_006714797.1 | XP_006714860.1 | 3 | 348 | |||
XM_011537684.2 | XP_011535986.1 | 2 | 254 | |||
XM_017009899.1 | XP_016865388.1 | 3 | 337 | |||
XM_017009900.1 | XP_016865389.1 | 2 | 254 | |||
XM_017009901.1 | XP_016865390.1 | 2 | 254 | |||
XM_017009902.1 | XP_016865391.1 | 2 | 254 | |||
GenBank mRNA | AK024024.1 | - | 3 | 279 | ||
AK308414.1 | - | 3 | 307 | |||
BC033454.1 | - | 3 | 292 | 4510274 | ||
BC050424.1 | - | 2 | 264 | 6472693 | ||
BI459318.1 | - | 4 | 400 | |||
DA399611.1 | - | 3 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
