siRNA Details
Gene Symbol: CDH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252338.2 | NP_001239267.1 | 15 | 5164 | ||
NM_001252339.2 | NP_001239268.1 | 15 | 5321 | |||
NM_001794.4 | NP_001785.2 | 16 | 5349 | |||
GenBank mRNA | AK025855.1 | - | 1 | 527 | ||
AK092092.1 | - | 1 | 1330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
