siRNA Details
Gene Symbol: NUP210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024923.3 | NP_079199.2 | 4 | 607 | ||
XM_005264994.3 | XP_005265051.1 | 4 | 607 | |||
XM_011533538.2 | XP_011531840.1 | 4 | 607 | |||
XM_017006021.1 | XP_016861510.1 | 4 | 607 | |||
GenBank mRNA | BC067089.1 | - | 4 | 525 | 6281693 |
