siRNA Details
Gene Symbol: WWC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024949.5 | NP_079225.5 | 16 | 2635 | ||
XM_011532269.2 | XP_011530571.1 | 16 | 3268 | |||
GenBank mRNA | AJ566366.1 | - | 3 | 317 | ||
AK074260.1 | - | 3 | 347 | |||
AK126057.1 | - | 17 | 2529 | |||
AK127061.1 | - | 9 | 1547 | |||
BC107683.1 | - | 7 | 1232 | |||
BX647704.1 | - | 4 | 362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
