siRNA Details
Gene Symbol: THNSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024838.4 | NP_079114.3 | 3 | 3510 | ||
XM_005252597.2 | XP_005252654.1 | 3 | 3586 | |||
XM_017016665.1 | XP_016872154.1 | 4 | 3596 | |||
GenBank mRNA | AK025655.1 | - | 1 | 1734 | ||
BX647989.1 | - | 3 | 3423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
