siRNA Details
Gene Symbol: JADE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287441.1 | NP_001274370.1 | 9 | 1798 | ||
NM_024900.4 | NP_079176.2 | 9 | 1919 | |||
GenBank mRNA | AF520952.1 | - | 9 | 1816 | ||
AK027620.1 | - | 9 | 1839 | |||
AK074986.1 | - | 4 | 1146 | |||
BC032376.1 | - | 9 | 1753 | 5111727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
