siRNA Details
Gene Symbol: ABCA11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002451.2 | 3 | 1398 | |||
GenBank mRNA | AK024359.1 | - | 3 | 1318 | ||
BC098116.1 | - | 4 | 1306 | |||
BC098259.1 | - | 4 | 1306 | |||
BC099730.1 | - | 3 | 1386 | |||
BC105084.1 | - | 2 | 1124 | |||
BC112069.1 | - | 2 | 1124 | |||
BC143677.1 | - | 3 | 1190 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
