siRNA Details
Gene Symbol: WLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY359035.1 | - | 12 | 2415 | ||
BC007211.1 | - | 11 | 1971 | |||
BC110826.1 | - | 12 | 2334 | 6183768 | ||
BC137109.1 | - | 12 | 2230 | 9021106 | ||
BX648748.1 | - | 11 | 2103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
