siRNA Details
Gene Symbol: ZCCHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024936.2 | NP_079212.2 | 13 | 2401 | ||
XM_011513835.1 | XP_011512137.1 | 14 | 2413 | |||
XM_017008128.1 | XP_016863617.1 | 12 | 2280 | |||
XM_017008129.1 | XP_016863618.1 | 11 | 2116 | |||
GenBank mRNA | AK026677.1 | - | 8 | 1670 | ||
AY629351.1 | - | 13 | 2394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
