siRNA Details
Gene Symbol: FRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166133.1 | NP_001159605.1 | 37 | 5299 | ||
NM_025074.6 | NP_079350.5 | 37 | 5299 | |||
XM_006714316.2 | XP_006714379.1 | 37 | 5546 | |||
GenBank mRNA | AK025684.1 | - | 4 | 207 | ||
BC131820.1 | - | 37 | 4889 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
