siRNA Details
Gene Symbol: ANP32A-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026808.1 | 1 | 725 | |||
GenBank mRNA | AK021784.1 | - | 1 | 725 | ||
BC132845.1 | - | 1 | 385 | |||
BC136847.1 | - | 1 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
