siRNA Details
Gene Symbol: FAM172A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005272111.4 | XP_005272168.1 | 8 | 3981 | ||
XM_017009962.1 | XP_016865451.1 | 7 | 3847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
