siRNA Details
Gene Symbol: ERCC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022657.1 | - | 1 | 512 | ||
AK226129.1 | - | 7 | 1285 | |||
BC093802.1 | - | 1 | 109 | |||
BC112227.1 | - | 1 | 109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
