siRNA Details
Gene Symbol: GVINP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003945.1 | 1 | 6151 | |||
GenBank mRNA | AK023435.1 | - | 1 | 736 | ||
AL834507.1 | - | 1 | 802 | |||
BX538342.1 | - | 1 | 2253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
