siRNA Details
Gene Symbol: LOC107983947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510285.1 | XP_011508587.1 | 1 | 604 | ||
GenBank mRNA | BC112311.1 | - | 1 | 646 | ||
BC112313.1 | - | 1 | 646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
