siRNA Details
Gene Symbol: ADGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025048.3 | NP_079324.2 | 8 | 1213 | ||
GenBank mRNA | AK026337.1 | - | 8 | 1144 | ||
AK225460.1 | - | 8 | 1144 | |||
BC104214.1 | - | 7 | 1062 | |||
BC104215.2 | - | 5 | 1183 | |||
CR627234.1 | - | 7 | 1050 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
