siRNA Details
Gene Symbol: MROH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163629.1 | NP_001157101.1 | 7 | 606 | ||
NM_025063.2 | NP_079339.2 | 7 | 582 | |||
XM_011510005.2 | XP_011508307.1 | 7 | 606 | |||
XM_011510006.2 | XP_011508308.1 | 7 | 606 | |||
GenBank mRNA | AK027203.1 | - | 7 | 550 | ||
BC113390.1 | - | 7 | 537 | |||
BC113392.1 | - | 7 | 537 | |||
DB230154.1 | - | 7 | 573 |
