siRNA Details
Gene Symbol: LOC100507388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121617.1 | 3 | 896 | |||
GenBank mRNA | AK027257.1 | - | 3 | 897 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
