siRNA Details
Gene Symbol: FRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166133.1 | NP_001159605.1 | 42 | 6449 | ||
GenBank mRNA | AK025684.1 | - | 9 | 1358 | ||
AL831853.1 | - | 3 | 648 | |||
BC131820.1 | - | 42 | 6039 | |||
BC131821.1 | - | 10 | 2636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
