siRNA Details
Gene Symbol: OPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025136.3 | NP_079412.1 | 2 | 750 | ||
XM_006723403.3 | XP_006723466.1 | 3 | 712 | |||
GenBank mRNA | AK025840.1 | - | 2 | 705 | ||
AK296429.1 | - | 2 | 626 | |||
BC005059.1 | - | 2 | 671 | 2957964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
