siRNA Details
Gene Symbol: RUNX1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026812.1 | 1 | 750 | |||
GenBank mRNA | AK024509.1 | - | 1 | 750 | ||
BC117305.1 | - | 1 | 190 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
