siRNA Details
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002814.2 | NP_001002814.2 | 1, 2 | 426 | ||
NM_025151.4 | NP_079427.4 | 1, 2 | 426 | |||
XM_017013869.1 | XP_016869358.1 | 1, 2 | 445 | |||
GenBank mRNA | AF368294.1 | - | 1, 2 | 396 | ||
AK226111.1 | - | 1, 2 | 116 | |||
AK291781.1 | - | 1, 2 | 423 | |||
AY280968.1 | - | 1, 2 | 383 | |||
BC077720.1 | - | 1, 2 | 396 | 6537110 | ||
DQ236342.1 | - | 1, 2 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
