siRNA Details
Gene Symbol: OGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029908.1 | XP_016885397.1 | 4 | 704 | ||
GenBank mRNA | AK300267.1 | - | 4 | 735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
