siRNA Details
Gene Symbol: GON4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282856.1 | NP_001269785.1 | 7, 8 | 1181 | ||
NM_001282858.1 | NP_001269787.1 | 7, 8 | 1233 | |||
NM_001282860.1 | NP_001269789.1 | 7, 8 | 1233 | |||
NM_001282861.1 | NP_001269790.1 | 7, 8 | 1233 | |||
NM_032292.5 | NP_115668.4 | 7, 8 | 1221 | |||
XM_005245284.3 | XP_005245341.1 | 7, 8 | 1173 | |||
XM_006711393.3 | XP_006711456.1 | 7, 8 | 1192 | |||
XM_006711394.3 | XP_006711457.1 | 7, 8 | 1178 | |||
XM_011509658.2 | XP_011507960.1 | 7, 8 | 1160 | |||
XM_011509659.2 | XP_011507961.1 | 7, 8 | 1197 | |||
GenBank mRNA | AB195687.1 | - | 7, 8 | 1233 | ||
AB195688.1 | - | 7, 8 | 1221 | |||
AB232668.1 | - | 7, 8 | 1109 | |||
AK026693.1 | - | 3, 4 | 142 | |||
AY335490.2 | - | 7, 8 | 1136 | |||
AY335491.2 | - | 7, 8 | 1181 | |||
AY335492.2 | - | 7, 8 | 1107 | |||
BC052260.1 | - | 5, 6 | 399 | |||
BC114629.1 | - | 7, 8 | 1108 | 40082464 | ||
BC115703.1 | - | 7, 8 | 1120 | 40082468 | ||
BX537764.1 | - | 7, 8 | 1109 | |||
BX648043.1 | - | 7, 8 | 1132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
