siRNA Details
Gene Symbol: GRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002086.4 | NP_002077.1 | 6 | 2755 | ||
NM_203506.2 | NP_987102.1 | 5 | 2633 | |||
GenBank mRNA | AF171699.1 | - | 1 | 333 | ||
AF246238.1 | - | 1 | 704 | |||
AF302079.1 | - | 1 | 323 | |||
AK024539.1 | - | 1 | 677 | |||
BC019082.2 | - | 1 | 716 | |||
BC033644.1 | - | 1 | 656 | |||
BC043007.1 | - | 1 | 581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
