siRNA Details
Gene Symbol: DCAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163484.1 | NP_001156956.1 | 3 | 469 | ||
NM_025230.4 | NP_079506.3 | 3 | 919 | |||
NR_028099.1 | 3 | 896 | ||||
GenBank mRNA | AB209073.1 | - | 3 | 882 | ||
AF130070.1 | - | 2 | 179 | |||
AF267858.1 | - | 3 | 582 | |||
AK074296.1 | - | 3 | 884 | |||
AK307849.1 | - | 3 | 354 | |||
BC067132.1 | - | 3 | 342 | 6170335 | ||
BX247970.1 | - | 2 | 410 | |||
BX648224.1 | - | 3 | 401 | |||
DB003266.1 | - | 2 | 501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
