siRNA Details
Gene Symbol: MYO19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163735.1 | NP_001157207.1 | 22 | 3226 | ||
NM_025109.5 | NP_079385.2 | 18 | 2626 | |||
XM_011525286.1 | XP_011523588.1 | 25 | 2913 | |||
XM_011525287.1 | XP_011523589.1 | 25 | 2907 | |||
XM_011525290.2 | XP_011523592.1 | 23 | 2775 | |||
XM_017025157.1 | XP_016880646.1 | 25 | 2887 | |||
XM_017025158.1 | XP_016880647.1 | 24 | 2820 | |||
XM_017025159.1 | XP_016880648.1 | 23 | 2781 | |||
XM_017025160.1 | XP_016880649.1 | 25 | 2913 | |||
XM_017025161.1 | XP_016880650.1 | 23 | 2705 | |||
GenBank mRNA | AB209035.1 | - | 23 | 4451 | ||
AK304073.1 | - | 22 | 2577 | |||
AL133017.1 | - | 9 | 3051 | |||
BC008900.2 | - | 18 | 2017 | 3629870 | ||
BU543266.1 | - | 4 | 367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
