siRNA Details
Gene Symbol: DCAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163484.1 | NP_001156956.1 | 3 | 472 | ||
NM_025230.4 | NP_079506.3 | 3 | 922 | |||
NR_028099.1 | 3 | 899 | ||||
GenBank mRNA | AB209073.1 | - | 3 | 885 | ||
AF130070.1 | - | 2 | 182 | |||
AF267858.1 | - | 3 | 585 | |||
AK074296.1 | - | 3 | 887 | |||
AK307849.1 | - | 3 | 357 | |||
BC067132.1 | - | 3 | 345 | 6170335 | ||
BX247970.1 | - | 2 | 413 | |||
BX648224.1 | - | 3 | 404 | |||
DB003266.1 | - | 2 | 504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
