siRNA Details
Gene Symbol: ITIH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001851.2 | NP_001001851.1 | 2 | 224 | ||
NM_030569.6 | NP_085046.5 | 2 | 224 | |||
XM_011519713.2 | XP_011518015.1 | 2 | 224 | |||
XM_011519714.2 | XP_011518016.1 | 2 | 224 | |||
GenBank mRNA | AK027375.1 | - | 2 | 161 | ||
AK075381.1 | - | 2 | 197 | |||
AL833203.1 | - | 2 | 69 | |||
AY238437.1 | - | 2 | 120 | |||
AY358426.1 | - | 2 | 189 | |||
BC136439.1 | - | 2 | 197 | |||
DA246953.1 | - | 2 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
