siRNA Details
Gene Symbol: B3GNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030765.2 | NP_110392.1 | 3 | 563 | ||
XM_011538732.1 | XP_011537034.1 | 2 | 400 | |||
GenBank mRNA | AB049586.1 | - | 2 | 433 | ||
AJ130848.1 | - | 1 | 270 | |||
AJ278962.1 | - | 1 | 270 | |||
AK095746.1 | - | 2 | 400 | |||
AL834452.1 | - | 2 | 377 | |||
AY358940.1 | - | 3 | 701 | |||
BC031399.1 | - | 3 | 530 | 5197310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
