siRNA Details
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317950.1 | NP_001304879.1 | 11 | 2688 | ||
NM_001317952.1 | NP_001304881.1 | 10 | 2246 | |||
NM_030767.5 | NP_110394.3 | 11 | 2633 | |||
XM_005252244.2 | XP_005252301.1 | 13 | 2805 | |||
XM_005252245.1 | XP_005252302.1 | 12 | 2663 | |||
XM_005252247.4 | XP_005252304.1 | 11 | 2684 | |||
XM_005252248.2 | XP_005252305.1 | 11 | 2801 | |||
XM_006717294.1 | XP_006717357.1 | 11 | 2617 | |||
XM_006717295.2 | XP_006717358.1 | 11 | 2801 | |||
XM_011519063.2 | XP_011517365.1 | 10 | 2296 | |||
XM_011519064.2 | XP_011517366.1 | 11 | 2801 | |||
XM_011519065.2 | XP_011517367.2 | 11 | 2276 | |||
XM_011519066.2 | XP_011517368.1 | 11 | 2801 | |||
XM_017015172.1 | XP_016870661.1 | 12 | 2403 | |||
GenBank mRNA | AB075848.1 | - | 11 | 2806 | ||
AF286341.1 | - | 1 | 188 | |||
AK024431.1 | - | 9 | 1581 | |||
AK074040.1 | - | 10 | 1676 | |||
AK131082.2 | - | 11 | 2688 | |||
AK160382.1 | - | 11 | 2689 | |||
AL833220.1 | - | 8 | 1110 | |||
AY703039.1 | - | 12 | 2288 | |||
AY703040.1 | - | 10 | 2246 | |||
AY703041.1 | - | 12 | 2002 | |||
AY703042.1 | - | 10 | 1815 | |||
AY703043.1 | - | 10 | 2606 | |||
AY703044.1 | - | 9 | 2300 | |||
AY703045.1 | - | 2 | 383 | |||
BC055285.1 | - | 11 | 2633 | 6143155 |
