siRNA Details
Gene Symbol: CYFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037333.2 | NP_001032410.1 | 31 | 5515 | ||
NM_001291721.1 | NP_001278650.1 | 30 | 5437 | |||
NM_001291722.1 | NP_001278651.1 | 32 | 5590 | |||
NM_014376.3 | NP_055191.2 | 31 | 5641 | |||
GenBank mRNA | AF132197.1 | - | 1 | 651 | ||
AK172724.1 | - | 1 | 1406 | |||
BC026892.2 | - | 1 | 337 | |||
BC066933.1 | - | 1 | 838 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
