siRNA Details
Gene Symbol: FZD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003468.3 | NP_003459.2 | 2 | 5027 | ||
GenBank mRNA | AK000125.1 | - | 1 | 230 | ||
AK024850.1 | - | 1 | 938 | |||
AL136852.1 | - | 1 | 570 | |||
BC095474.1 | - | 1 | 832 | |||
CR533442.1 | - | 1 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
