siRNA Details
Gene Symbol: MIR600HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026677.1 | 1 | 4681 | |||
GenBank mRNA | AF251293.1 | - | 1 | 370 | ||
AK025814.1 | - | 1 | 1843 | |||
AK123844.1 | - | 1 | 2411 | |||
BC015678.2 | - | 1 | 2671 | |||
BC034918.1 | - | 1 | 633 | |||
BX648949.1 | - | 1 | 4328 | |||
CR749230.1 | - | 1 | 4686 |
