siRNA Details
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317950.1 | NP_001304879.1 | 21 | 4121 | ||
NM_001317952.1 | NP_001304881.1 | 20 | 3679 | |||
NM_030767.5 | NP_110394.3 | 21 | 4066 | |||
XM_005252244.2 | XP_005252301.1 | 23 | 4238 | |||
XM_005252245.1 | XP_005252302.1 | 22 | 4096 | |||
XM_005252247.4 | XP_005252304.1 | 21 | 4117 | |||
XM_005252248.2 | XP_005252305.1 | 20 | 4138 | |||
XM_006717294.1 | XP_006717357.1 | 21 | 4050 | |||
XM_011519063.2 | XP_011517365.1 | 20 | 3729 | |||
XM_011519065.2 | XP_011517367.2 | 21 | 3709 | |||
XM_017015172.1 | XP_016870661.1 | 22 | 3836 | |||
GenBank mRNA | AF286341.1 | - | 11 | 1620 | ||
AK131082.2 | - | 21 | 4121 | |||
AK160382.1 | - | 21 | 4123 | |||
AY703039.1 | - | 22 | 3721 | |||
AY703040.1 | - | 20 | 3679 | |||
AY703043.1 | - | 20 | 4039 | |||
AY703044.1 | - | 19 | 3733 | |||
AY703045.1 | - | 12 | 1812 | |||
AY703046.1 | - | 8 | 938 | |||
BC042202.1 | - | 7 | 796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
