siRNA Details
Gene Symbol: FBXO38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271723.1 | NP_001258652.1 | 15, 16 | 2195 | ||
NM_030793.4 | NP_110420.3 | 16, 17 | 2705 | |||
XM_005268513.1 | XP_005268570.1 | 16, 17 | 2919 | |||
XM_006714797.1 | XP_006714860.1 | 16, 17 | 2793 | |||
XM_011537684.2 | XP_011535986.1 | 16, 17 | 2972 | |||
XM_017009899.1 | XP_016865388.1 | 17, 18 | 3067 | |||
XM_017009900.1 | XP_016865389.1 | 17, 18 | 3131 | |||
XM_017009901.1 | XP_016865390.1 | 16, 17 | 2759 | |||
XM_017009902.1 | XP_016865391.1 | 16, 17 | 2747 | |||
GenBank mRNA | AF177339.1 | - | 2, 3 | 851 | ||
AF251055.1 | - | 9, 10 | 1643 | |||
AK308414.1 | - | 15, 16 | 2144 | |||
AK313558.1 | - | 3, 4 | 1009 | |||
AL832841.1 | - | 6, 7 | 1397 | |||
BC005849.2 | - | 6, 7 | 1504 | |||
BC005873.2 | - | 6, 7 | 1504 | |||
BC015366.1 | - | 2, 3 | 576 | |||
BC033454.1 | - | 16, 17 | 2638 | 4510274 | ||
BC050424.1 | - | 14, 15 | 2100 | 6472693 | ||
BC056147.1 | - | 2, 3 | 756 | |||
BC078159.1 | - | 2, 3 | 596 |

More Information
Other Species Targeted

Gene Ontology Categories:
