siRNA Details
Gene Symbol: C1orf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030806.3 | NP_110433.1 | 6 | 1471 | ||
GenBank mRNA | AF312864.1 | - | 6 | 1393 | ||
AK056487.1 | - | 6 | 1207 | |||
AY258286.1 | - | 6 | 1384 | |||
BC028567.2 | - | 6 | 1429 | 3635521 |
