siRNA Details
Gene Symbol: ACSBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ577571.1 | - | 5 | 645 | ||
AK225862.1 | - | 4 | 450 | |||
AK302604.1 | - | 5 | 699 | |||
AL136854.1 | - | 5 | 483 | |||
AY358766.1 | - | 5 | 742 | |||
BC022027.1 | - | 5 | 673 | 4825681 | ||
BC090046.1 | - | 5 | 658 | 30388900 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
