siRNA Details
Gene Symbol: CAB39L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079670.2 | NP_001073138.1 | 5, 6 | 670 | ||
NM_001287337.1 | NP_001274266.1 | 6, 7 | 808 | |||
NM_001287338.1 | NP_001274267.1 | 6, 7 | 803 | |||
NM_001287339.1 | NP_001274268.1 | 4, 5 | 472 | |||
NM_030925.3 | NP_112187.2 | 3, 4 | 721 | |||
XM_011535254.2 | XP_011533556.1 | 7, 8 | 1212 | |||
XM_011535255.2 | XP_011533557.1 | 5, 6 | 692 | |||
XM_011535256.2 | XP_011533558.1 | 6, 7 | 957 | |||
XM_017020785.1 | XP_016876274.1 | 6, 7 | 1084 | |||
XM_017020786.1 | XP_016876275.1 | 7, 8 | 1221 | |||
XM_017020787.1 | XP_016876276.1 | 7, 8 | 1474 | |||
XM_017020788.1 | XP_016876277.1 | 8, 9 | 1350 | |||
GenBank mRNA | AA278473.1 | - | 1, 2 | 157 | ||
AK022639.1 | - | 2, 3 | 266 | |||
AY211923.1 | - | 5, 6 | 583 | |||
AY288977.2 | - | 6, 7 | 735 | |||
DB457246.1 | - | 4, 5 | 472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
