siRNA Details
Gene Symbol: RAB1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030981.2 | NP_112243.1 | 5 | 490 | ||
XM_017018378.1 | XP_016873867.1 | 5 | 549 | |||
GenBank mRNA | AF092437.1 | - | 5 | 408 | ||
AJ245875.1 | - | 4 | 397 | |||
AK292086.1 | - | 5 | 458 | |||
AK315333.1 | - | 5 | 451 | |||
AL136635.1 | - | 5 | 442 | |||
BC028331.1 | - | 5 | 441 | |||
BC071169.1 | - | 5 | 429 | 6026121 | ||
CR533462.1 | - | 4 | 395 |
