siRNA Details
Gene Symbol: HYI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190880.2 | NP_001177809.1 | 3 | 546 | ||
NM_001243526.1 | NP_001230455.1 | 4 | 621 | |||
NM_031207.5 | NP_112484.3 | 3 | 546 | |||
XM_005271239.4 | XP_005271296.1 | 4 | 873 | |||
XM_005271240.4 | XP_005271297.1 | 3 | 771 | |||
XM_006710937.3 | XP_006711000.1 | 4 | 873 | |||
XM_011542222.2 | XP_011540524.1 | 3 | 771 | |||
GenBank mRNA | AF284750.1 | - | 3 | 421 | ||
AF284751.1 | - | 3 | 421 | |||
AK096244.1 | - | 1 | 1622 | |||
AY037165.1 | - | 3 | 520 | |||
AY775560.1 | - | 3 | 521 | |||
BC006140.1 | - | 3 | 232 | 3928690 | ||
BC019041.2 | - | 3 | 348 | 4586546 | ||
BM715679.1 | - | 3 | 519 | |||
BM826556.1 | - | 4 | 288 | |||
JF911821.1 | - | 2 | 131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
