siRNA Details
Gene Symbol: PRKY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028062.1 | 3 | 847 | |||
GenBank mRNA | BC074851.2 | - | 3 | 524 | 30915351 | |
BC074852.2 | - | 3 | 524 | 30915567 | ||
Y10485.1 | - | 2 | 246 | |||
Y15801.1 | - | 3 | 846 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
