siRNA Details
Gene Symbol: TBL1XR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321193.1 | NP_001308122.1 | 16 | 5797 | ||
NM_001321194.1 | NP_001308123.1 | 17 | 5977 | |||
NM_001321195.1 | NP_001308124.1 | 15 | 5694 | |||
NM_024665.5 | NP_078941.2 | 16 | 5858 | |||
XM_005247775.2 | XP_005247832.1 | 15 | 5813 | |||
XM_006713746.1 | XP_006713809.1 | 17 | 5867 | |||
XM_011513141.1 | XP_011511443.1 | 17 | 5801 | |||
XM_011513142.2 | XP_011511444.1 | 16 | 5782 | |||
XM_011513143.2 | XP_011511445.1 | 15 | 5708 | |||
XM_017007185.1 | XP_016862674.1 | 16 | 5795 | |||
GenBank mRNA | AF267864.1 | - | 1 | 856 | ||
AK025746.1 | - | 1 | 1508 | |||
BC034712.1 | - | 1 | 1029 | |||
BC048298.1 | - | 1 | 803 | |||
BC092400.1 | - | 1 | 1413 | |||
BX648080.1 | - | 2 | 5470 | |||
BX648935.1 | - | 16 | 5796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
