siRNA Details
Gene Symbol: ZNF142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105537.2 | NP_001099007.1 | 5 | 806 | ||
NR_073599.1 | 6 | 1115 | ||||
NR_073600.1 | 6 | 948 | ||||
XM_006712730.3 | XP_006712793.1 | 6 | 1481 | |||
XM_011511784.2 | XP_011510086.1 | 6 | 1576 | |||
XM_011511785.2 | XP_011510087.1 | 6 | 1501 | |||
XM_011511786.2 | XP_011510088.1 | 6 | 1406 | |||
XM_011511787.2 | XP_011510089.1 | 5 | 1401 | |||
XM_011511788.2 | XP_011510090.1 | 5 | 1306 | |||
XM_011511789.2 | XP_011510091.1 | 5 | 974 | |||
XM_017004871.1 | XP_016860360.1 | 5 | 1557 | |||
XM_017004872.1 | XP_016860361.1 | 4 | 957 | |||
GenBank mRNA | BC033631.1 | - | 5 | 1529 | ||
BC053849.2 | - | 6 | 1134 | |||
D87073.1 | - | 5 | 804 |
