siRNA Details
Gene Symbol: DRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524025.2 | XP_011522327.1 | 1 | 583 | ||
GenBank mRNA | AL136926.1 | - | 6 | 1278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
