siRNA Details
Gene Symbol: TBL1XR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321193.1 | NP_001308122.1 | 16 | 5488 | ||
NM_001321194.1 | NP_001308123.1 | 17 | 5668 | |||
NM_001321195.1 | NP_001308124.1 | 15 | 5385 | |||
NM_024665.5 | NP_078941.2 | 16 | 5549 | |||
XM_005247775.2 | XP_005247832.1 | 15 | 5504 | |||
XM_006713746.1 | XP_006713809.1 | 17 | 5558 | |||
XM_011513141.1 | XP_011511443.1 | 17 | 5492 | |||
XM_011513142.2 | XP_011511444.1 | 16 | 5473 | |||
XM_011513143.2 | XP_011511445.1 | 15 | 5399 | |||
XM_017007185.1 | XP_016862674.1 | 16 | 5486 | |||
GenBank mRNA | AF267864.1 | - | 1 | 547 | ||
AK025746.1 | - | 1 | 1199 | |||
BC034712.1 | - | 1 | 720 | |||
BC048298.1 | - | 1 | 494 | |||
BC092400.1 | - | 1 | 1104 | |||
BX648080.1 | - | 2 | 5161 | |||
BX648935.1 | - | 16 | 5487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
