siRNA Details
Gene Symbol: CAB39L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079670.2 | NP_001073138.1 | 11 | 2359 | ||
NM_001287337.1 | NP_001274266.1 | 12 | 2497 | |||
NM_001287338.1 | NP_001274267.1 | 12 | 2492 | |||
NM_001287339.1 | NP_001274268.1 | 10 | 2161 | |||
NM_030925.3 | NP_112187.2 | 9 | 2410 | |||
XM_011535254.2 | XP_011533556.1 | 13 | 2901 | |||
XM_011535255.2 | XP_011533557.1 | 11 | 2381 | |||
XM_011535256.2 | XP_011533558.1 | 12 | 2646 | |||
XM_017020785.1 | XP_016876274.1 | 12 | 2773 | |||
XM_017020786.1 | XP_016876275.1 | 13 | 2910 | |||
XM_017020787.1 | XP_016876276.1 | 13 | 3163 | |||
XM_017020788.1 | XP_016876277.1 | 14 | 3039 | |||
GenBank mRNA | AK022639.1 | - | 8 | 1955 | ||
BX647518.1 | - | 11 | 2296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
