siRNA Details
Gene Symbol: ZNF611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161499.1 | NP_001154971.1 | 6 | 3545 | ||
NM_001161500.1 | NP_001154972.1 | 5 | 3445 | |||
NM_001161501.1 | NP_001154973.1 | 5 | 3408 | |||
NM_030972.3 | NP_112234.3 | 7 | 3587 | |||
GenBank mRNA | AK021787.1 | - | 1 | 1330 | ||
AK091389.1 | - | 2 | 3126 | |||
AL834123.1 | - | 1 | 2949 | |||
BC000918.2 | - | 1 | 980 | |||
BC028590.1 | - | 9 | 3829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
